(2,5-Difluorophenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Difluorophenylacetonitrile, 2,5-Difluorobenzyl cyanide

005585-25G 005585-100G
101778-518EA 76.92 USD
101778-518 102832-770
(2,5-Difluorophenyl)acetonitrile ≥98%
(2,5-Difluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₅F₂N
MW: 153.13 g/mol
Density: 1.233
MDL Number: MFCD00009972
CAS Number: 69584-87-8
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.233
ND20 1.483

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR