(2,4-Difluorophenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

005584-5G 005584-25G
101778-512EA 15.38 USD
101778-512 101778-514
(2,4-Difluorophenyl)acetonitrile ≥98%
(2,4-Difluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₅F₂N
Density: 1.249
MDL Number: MFCD00009971
CAS Number: 656-35-9
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.249
ND20 1.48


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR