(2,3-Difluorophenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3-Difluorobenzyl cyanide, 2-(2,3-Difluorophenyl)acetonitrile, 2,3-Difluorophenylacetonitrile

005583-1G 005583-5G 005583-25G
101778-506EA 16.92 USD
101778-506 101778-508 101778-510
(2,3-Difluorophenyl)acetonitrile ≥98%
(2,3-Difluorophenyl)acetonitrile

Hazard: Toxic

Formula: C₈H₅F₂N
MW: 153.13 g/mol
MDL Number: MFCD00061277
CAS Number: 145689-34-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR