2,6-Difluorophenol ≥98%

Supplier: Matrix Scientific

005564-25G 005564-100G 005564-5G
101778-498EA 200 USD
101778-498 101778-500 101778-496
2,6-Difluorophenol ≥98%
2,6-Difluorophenol

Hazard: Flammable/Corrosive

Formula: C₆H₄F₂O
Boiling Pt: 59…61 °C (17 mmHg)
Melting Pt: 38…41 °C
Flash Pt: 58 °C
MDL Number: MFCD00002158
CAS Number: 28177-48-2
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Boiling point 59-61°/17m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR