2,4-Difluorophenol ≥99%

Supplier: Matrix Scientific

005562-25G 005562-100G
101778-492EA 53.85 USD
101778-492 101778-494
2,4-Difluorophenol ≥99%
2,4-Difluorophenol

Hazard: Flammable/Corrosive

Formula: C₆H₄F₂O
Boiling Pt: 52-53°C/19mm
Melting Pt: 22.4°C
Density: 1.362
Flash Pt: 56°C
MDL Number: MFCD00009715
CAS Number: 367-27-1
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Boiling point 52-53°/19m
Density 1.362
ND20 1.486


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR