2,5-Difluorophenol ≥95%

Supplier: Matrix Scientific

005561-25G 005561-100G
101778-488EA 73.85 USD
101778-488 101778-490
2,5-Difluorophenol ≥95%
2,5-Difluorophenol

Hazard: Flammable Solid/Corrosive

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Melting Pt: 40…42 °C
Flash Pt: 53 °C
MDL Number: MFCD00042501
CAS Number: 2713-31-7
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Melting point 40-42°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR