2,6-Dichlorophenethylamine ≥99%
Supplier: Matrix Scientific
Synonyms:
2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine
005559-1G
005559-5G
101778-484EA
50.77
USD
101778-484
101778-486
2,6-Dichlorophenethylamine ≥99%
2,6-Dichlorophenethylamine
Hazard: Corrosive/Irritant
Formula:
C₈H₉Cl₂N MW: 190.07 g/mol Boiling Pt: 90 °C (1 mmHg) Density: 1.28 |
MDL Number:
MFCD00060612 CAS Number: 14573-23-0 |
Specification Test Results
Boiling point | 90°/1mm |
Density | 1.28 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...