1,2-Dichlorooctafluorocyclohexene ≥97%

Supplier: Matrix Scientific

005548-5G 005548-1G
101778-478EA 287.69 USD
101778-478 101778-476
1,2-Dichlorooctafluorocyclohexene ≥97%
1,2-Dichlorooctafluorocyclohexene

Hazard: Toxic

Formula: C₆Cl₂F₈
Boiling Pt: 113 °C
Density: 1.719
MDL Number: MFCD00039466
CAS Number: 336-19-6
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 113°
Density 1.719
ND20 1.375


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR