4-((Trifluoromethyl)thio)benzoyl chloride ≥97%

Supplier: Matrix Scientific

005542-5G 005542-25G
101778-468EA 192.31 USD
101778-468 101778-470
4-((Trifluoromethyl)thio)benzoyl chloride ≥97%
4-((Trifluoromethyl)thio)benzoyl chloride

Hazard: Corrosive

Formula: C₈H₄ClF₃OS
MW: 240.63 g/mol
Density: 1.43
MDL Number: MFCD01631632
CAS Number: 330-14-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.43

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR