2-(Trifluoromethylthio)benzyl bromide
Supplier: Matrix Scientific
005531-250MG
005531-1G
101778-458EA
46.15
USD
101778-458
101778-460
2-(Trifluoromethylthio)benzyl bromide
2-(Trifluoromethylthio)benzyl bromide
Hazard: Corrosive
Formula:
C₈H₆BrF₃S MW: 271.10 Boiling Pt: 71°C/1 torr |
MDL Number:
MFCD00236345 CAS Number: 238403-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...