2-(Trifluoromethyl)phenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

Synonyms: α,α,α-Trifluoro-o-tolyl isothiocyanate

005514-1G 005514-5G
101778-454EA 21.54 USD
101778-454 101778-456
2-(Trifluoromethyl)phenyl isothiocyanate ≥98%
2-(Trifluoromethyl)phenyl isothiocyanate

Hazard: Toxic/Corrosive/Lachrymator

Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 96 °C (10 mmHg)
Density: 1.346
MDL Number: MFCD00039644
CAS Number: 1743-86-8

Order Now

Specification Test Results

Boiling point 96°/10mm
Density 1.346
ND20 1.566

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR