2-(Trifluoromethyl)phenylacetonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)benzyl cyanide, 1-(Cyanmethyl)-2-(trifluormethyl)benzene
005511-25G
005511-5G
101778-446EA
89.23
USD
101778-446
101778-444
2-(Trifluoromethyl)phenylacetonitrile ≥98%
2-(Trifluoromethyl)phenylacetonitrile
Hazard: Toxic
Formula:
C₉H₆F₃N MW: 185.15 g/mol Boiling Pt: 92 °C (11 mmHg) Melting Pt: 29…30 °C |
MDL Number:
MFCD00001903 CAS Number: 3038-47-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Boiling point | 92°/11mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...