α,α,α-Trifluoro-o-toluoyl chloride

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethyl)benzoyl chloride, 2-(Trifluoromethyl)benzene-1-carbonyl chloride

005509-100G 005509-25G
101778-442EA 113.85 USD
101778-442 101778-440
α,α,α-Trifluoro-o-toluoyl chloride
α,α,α-Trifluoro-o-toluoyl chloride

Hazard: Corrosive/Lachrymator

Formula: C₈H₄ClF₃O
Boiling Pt: 90 °C (16 mmHg)
Density: 1.416
MDL Number: MFCD00000667
CAS Number: 312-94-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 90°/16mm
Density 1.416
ND20 1.481


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR