3-(Trifluoromethyl)-3-hydroxybutyric acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4,4,4-Trifluoro-3-hydroxy-3-methylbutanoic acid, 3-Hydroxy-3-(trifluoromethyl)butyric acid

005507-5G 005507-25G
101778-432EA 130.77 USD
101778-432 101778-434
3-(Trifluoromethyl)-3-hydroxybutyric acid ≥95%
3-(Trifluoromethyl)-3-hydroxybutyric acid

Hazard: Corrosive/Stench

Formula: C₅H₇F₃O₃
MW: 172.1 g/mol
Boiling Pt: 87-90°C/0.5mm
Melting Pt: 39…41 °C
Density: 1.388
MDL Number: MFCD00039531
CAS Number: 338-03-4

Order Now

Specification Test Results

Boiling point 87-90°/0.5
Density 1.388

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR