4-(Trifluoromethyl)benzylamine ≥97%

Supplier: Matrix Scientific

005502-25G 005502-100G
101778-418EA 80 USD
101778-418 101778-420
4-(Trifluoromethyl)benzylamine ≥97%
4-(Trifluoromethyl)benzylamine

Hazard: Corrosive

Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 80-82°C/15mm
Density: 1.229
MDL Number: MFCD00010220
CAS Number: 3300-51-4

Order Now

Specification Test Results

Boiling point 80-82°/15m
Density 1.229
ND20 1.464

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR