α,α,α-Trifluoro-p-toluonitrile ≥98%

Supplier: Matrix Scientific

005501-25G 005501-100G 005501-500G
101778-412EA 18.46 USD
101778-412 101778-414 101778-416
α,α,α-Trifluoro-p-toluonitrile ≥98%
α,α,α-Trifluoro-p-toluonitrile

TCA listed: T
Hazard: Toxic/Lachrymator

Formula: C₈H₄F₃N
Boiling Pt: 80…81 °C (20 mmHg)
Melting Pt: 39…41 °C
Density: 1.278
MDL Number: MFCD00001826
CAS Number: 455-18-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 80-81°/20m
Density 1.278


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR