α'-Bromo-α,α,α-trifluoro-o-xylene ≥98%
Supplier: Matrix Scientific
005495-100G
005495-5G
005495-25G
101778-408EA
243.08
USD
101778-408
101778-404
101778-406
α'-Bromo-α,α,α-trifluoro-o-xylene ≥98%
α'-Bromo-α,α,α-trifluoro-o-xylene
Hazard: Corrosive/Irritant/Lachrymator
Formula:
C₈H₆BrF₃ Boiling Pt: 70…72 °C (7 mmHg) Melting Pt: 34…35 °C Density: 1.571 |
MDL Number:
MFCD00013558 CAS Number: 395-44-8 UN: 3261 ADR: 8,II |
Specification Test Results
Boiling point | 70-72°/7mm |
Density | 1.571 |
ND20 | 1.494 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...