3-(Trifluoromethyl)cyclohexanone ≥97%
Supplier: Matrix Scientific
005485-1G
005485-5G
101778-388EA
158.46
USD
101778-388
101778-390
3-(Trifluoromethyl)cyclohexanone ≥97%
3-(Trifluoromethyl)cyclohexanone
Hazard: Flammable/Irritant
Formula:
C₇H₉F₃O Boiling Pt: 61…62 °C (20 mmHg) |
MDL Number:
MFCD00034662 CAS Number: 585-36-4 UN: 1224 ADR: 3,III |
Specification Test Results
Boiling point | 61-62°/20m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...