2-(Trifluoromethyl)propene ≥97%

Supplier: Matrix Scientific

005483-25G 005483-5G
101778-386EA 846.15 USD
101778-386 101778-384
2-(Trifluoromethyl)propene ≥97%
2-(Trifluoromethyl)propene

Hazard: Flammable Gas

Formula: C₄H₅F₃
MW: 110.08 g/mol
Boiling Pt: 6…7 °C
MDL Number: MFCD00042093
CAS Number: 374-00-5

Order Now

Specification Test Results

Boiling point 6-7°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR