3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3,3,3-Trifluoro-2-hydroxyisobutyric Acid, 3,3,3-Trifluoro-2-hydroxy-2-methylpropanoic acid, 2-Hydroxy-2-(trifluoromethyl)propionic Acid
005472-5G
005472-25G
101778-374EA
110.77
USD
101778-374
101778-376
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥95%
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid
Hazard: Corrosive/Irritant
Formula:
C₄H₅F₃O₃ MW: 158.08 g/mol Boiling Pt: 189°C Melting Pt: 85 °C |
MDL Number:
MFCD00190646 CAS Number: 374-35-6 |
Specification Test Results
Boiling point | 189° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...