3-(Trifluoromethyl)anisole ≥99%
Supplier: Matrix Scientific
005467-25G
005467-100G
101778-370EA
21.54
USD
101778-370
101778-372
3-(Trifluoromethyl)anisole ≥99%
3-(Trifluoromethyl)anisole
Hazard: Flammable
Formula:
C₈H₇F₃O MW: 176.14 g/mol Boiling Pt: 160°C Density: 1.217 |
MDL Number:
MFCD00000385 CAS Number: 454-90-0 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 160° |
Density | 1.217 |
ND20 | 1.4434 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...