2-(Trifluoromethyl)benzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

005464-25G 005464-100G
101778-366EA 118.46 USD
101778-366 101778-368
2-(Trifluoromethyl)benzenesulfonyl chloride ≥98%
2-(Trifluoromethyl)benzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₄ClF₃O₂S
Boiling Pt: 136 °C (15 mmHg)
Density: 1.585
MDL Number: MFCD00051696
CAS Number: 776-04-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 136°/15mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR