4-(Trifluoromethyl)phenylacetonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(Trifluoromethyl)benzyl cyanide
005463-5G
005463-25G
005463-100G
101778-358EA
15.38
USD
101778-358
101778-360
101778-362
4-(Trifluoromethyl)phenylacetonitrile ≥97%
4-(Trifluoromethyl)phenylacetonitrile
Hazard: Toxic/Irritant
Formula:
C₉H₆F₃N MW: 185.15 g/mol Boiling Pt: 131…132 °C (20 mmHg) Melting Pt: 45…47 °C |
MDL Number:
MFCD00001921 CAS Number: 2338-75-2 UN: 3439 ADR: 6.1,III |
Specification Test Results
Boiling point | 131-132°/2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...