3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid

Supplier: Matrix Scientific

Synonyms: 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid

005462-5G 005462-25G
101778-352EA 130.77 USD
101778-352 101778-354
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid

Hazard: Corrosive/Refrigerate

Formula: C₄H₂F₆O₂
MW: 196.048 g/mol
Boiling Pt: 125°C/750mm
Melting Pt: 49.5…50.5 °C
MDL Number: MFCD00464165
CAS Number: 564-10-3

Order Now

Specification Test Results

Boiling point 125°/750mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR