4-(Trifluoromethyl)benzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

005458-25G 005458-100G
101778-348EA 49.23 USD
101778-348 101778-350
4-(Trifluoromethyl)benzenesulfonyl chloride ≥98%
4-(Trifluoromethyl)benzenesulphonyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₇H₄ClF₃O₂S
Boiling Pt: 118-120°C/18mm
Melting Pt: 27-30°C
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 118-120°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR