α'-Chloro-α,α,α-trifluoro-o-xylene ≥97%

Supplier: Matrix Scientific

005396-5G 005396-25G 005396-100G
101778-332EA 61.54 USD
101778-332 101778-334 101778-336
α'-Chloro-α,α,α-trifluoro-o-xylene ≥97%
α'-Chloro-α,α,α-trifluoro-o-xylene

Hazard: Corrosive/Lachrymator

Formula: C₈H₆ClF₃
Density: 1.337
MDL Number: MFCD00009920
CAS Number: 21742-00-7
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Density 1.337
ND20 1.469


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR