3-(Trifluoromethyl)benzylamine ≥98%

Supplier: Matrix Scientific

005395-5G 005395-25G
101778-328EA 41.54 USD
101778-328 101778-330
3-(Trifluoromethyl)benzylamine ≥98%
3-(Trifluoromethyl)benzylamine

Hazard: Corrosive

Formula: C₈H₈F₃N
MW: 175.15 g/mol
Density: 1.222
MDL Number: MFCD00008117
CAS Number: 2740-83-2

Order Now

Specification Test Results

Density 1.222
ND20 1.463

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR