4-(Trifluoromethoxy)benzoyl chloride ≥97%

Supplier: Matrix Scientific

005372-25G 005372-100G
101778-314EA 75.38 USD
101778-314 102832-766
4-(Trifluoromethoxy)benzoyl chloride ≥97%
4-(Trifluoromethoxy)benzoyl chloride

Hazard: Corrosive

Formula: C₈H₄ClF₃O₂
Boiling Pt: 90…92 °C (15 mmHg)
Density: 1.425
MDL Number: MFCD00052329
CAS Number: 36823-88-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 90-92°/15m
Density 1.425
ND20 1.472


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR