3-(Trifluoromethoxy)aniline ≥97%

Supplier: Matrix Scientific

005360-25G 005360-100G
101778-296EA 53.85 USD
101778-296 101778-298
3-(Trifluoromethoxy)aniline ≥97%
3-(Trifluoromethoxy)aniline

Hazard: Flammable/Irritant

Formula: C₇H₆F₃NO
Boiling Pt: 72…75 °C (8 mmHg)
Density: 1.325
MDL Number: MFCD00041511
CAS Number: 1535-73-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 72-75°/8mm
Density 1.325
ND20 1.4649


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR