4-(Trifluoromethoxy)benzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (4-(Trifluoromethoxy)phenyl)methanamine

005355-25G 005355-100G
101778-288EA 104.62 USD
101778-288 101778-290
4-(Trifluoromethoxy)benzylamine ≥95%
4-(Trifluoromethoxy)benzylamine

Hazard: Corrosive/Irritant

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 57-60°C/10mm
Density: 1.252
MDL Number: MFCD00061237
CAS Number: 93919-56-3

Order Now

Specification Test Results

Boiling point 57-60°/10m
Density 1.252
ND20 1.452

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR