4-(Trifluoromethoxy)benzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

005346-5G 005346-25G
101778-284EA 38.46 USD
101778-284 101778-286
4-(Trifluoromethoxy)benzenesulfonyl chloride ≥98%
4-(Trifluoromethoxy)benzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 120-122°C/15mm
Density: 1.536
MDL Number: MFCD00042408
CAS Number: 94108-56-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 120-122°/1
Density 1.536
ND20 1.479

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR