2-(Trifluoromethoxy)phenylacetonitrile
Supplier: Matrix Scientific
Synonyms:
1-(Cyanmethyl)-2-(trifluormethoxy)benzene, 2-(Trifluoromethoxy)benzyl cyanide
005330-25G
005330-5G
101778-272EA
90.77
USD
101778-272
101778-270
2-(Trifluoromethoxy)phenylacetonitrile
2-(Trifluoromethoxy)phenylacetonitrile
Hazard: Toxic
Formula:
C₉H₆F₃NO MW: 201.15 Boiling Pt: 222…223 °C Density: 1.28 Flash Pt: >110 °C |
MDL Number:
MFCD00236326 CAS Number: 137218-25-8 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...