2-(Trifluoromethoxy)phenol ≥97%

Supplier: Matrix Scientific

005328-5G 005328-25G
101778-266EA 21.54 USD
101778-266 101778-268
2-(Trifluoromethoxy)phenol ≥97%
2-(Trifluoromethoxy)phenol

Hazard: Irritant

Formula: C₇H₅F₃O₂
Boiling Pt: 69…71 °C (60 Torr)
Density: 1.332
MDL Number: MFCD00236324
CAS Number: 32858-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR