α-Bromo-2-(trifluoromethoxy)toluene ≥98%

Supplier: Matrix Scientific

005326-25G 005326-5G
101778-260EA 95.38 USD
101778-260 101778-258
α-Bromo-2-(trifluoromethoxy)toluene ≥98%
α-Bromo-2-(trifluoromethoxy)toluene

Hazard: Corrosive/Lachrymator

Formula: C₈H₆BrF₃O
MDL Number: MFCD00236321
CAS Number: 198649-68-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR