4-(Trifluoromethoxy)phenylacetonitrile ≥98%

Supplier: Matrix Scientific

005322-100G 005322-5G 005322-25G
101778-256EA 176.92 USD
101778-256 101778-252 101778-254
4-(Trifluoromethoxy)phenylacetonitrile ≥98%
4-(Trifluoromethoxy)phenylacetonitrile

Hazard: Toxic/Irritant-Harmful

Formula: C₉H₆F₃NO
Melting Pt: 30…33 °C
MDL Number: MFCD00061240
CAS Number: 49561-96-8
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 30-33°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR