3-(Trifluoromethoxy)phenylacetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethoxy)benzyl cyanide, 1-(Cyanmethyl)-3-(trifluormethoxy)benzol, (3-Trifluoromethoxyphenyl)acetonitrile

005321-5G 005321-1G 005321-25G
101778-248EA 104.62 USD
101778-248 101778-246 101778-250
3-(Trifluoromethoxy)phenylacetonitrile ≥97%
3-(Trifluoromethoxy)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₆F₃NO
MW: 201.15 g/mol
MDL Number: MFCD00061272
CAS Number: 108307-56-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR