2-Bromo-4'-(trifluoromethoxy)acetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-(4-(trifluoromethoxy)phenyl)ethanone, 4-(Trifluoromethoxy)phenacyl bromide

005320-5G 005320-25G 005320-100G
101778-240EA 41.54 USD
101778-240 101778-242 101778-244
2-Bromo-4'-(trifluoromethoxy)acetophenone ≥97%
2-Bromo-4'-(trifluoromethoxy)acetophenone

Hazard: Corrosive

Formula: C₉H₆BrF₃O₂
MW: 283.04 g/mol
Melting Pt: 46-48°C
MDL Number: MFCD00052333
CAS Number: 103962-10-3

Order Now

Specification Test Results

Melting point 46-48°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR