3-(Trifluoromethoxy)benzylamine ≥97%

Supplier: Matrix Scientific

005319-1G 005319-25G
101778-234EA 13.85 USD
101778-234 101778-238
3-(Trifluoromethoxy)benzylamine ≥97%
3-(Trifluoromethoxy)benzylamine

Hazard: Corrosive

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD00061267
CAS Number: 93071-75-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR