4-(Trifluoromethoxy)benzoyl acetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Oxo-3-(4-(trifluoromethoxy)phenyl)propanenitrile

005317-5G 005317-1G
101778-228EA 1153.85 USD
101778-228 101778-226
4-(Trifluoromethoxy)benzoyl acetonitrile ≥97%
4-(Trifluoromethoxy)benzoyl acetonitrile

Hazard: Toxic

Formula: C₁₀H₆F₃NO₂
MW: 229.16 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD00052146
CAS Number: 122454-46-0

Order Now

Specification Test Results

Melting point 82-84°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR