α-Bromo-3-(trifluoromethoxy)toluene ≥97%
Supplier: Matrix Scientific
005302-1G
005302-5G
005302-25G
101778-216EA
20
USD
101778-216
101778-218
101778-220
α-Bromo-3-(trifluoromethoxy)toluene ≥97%
α-Bromo-3-(trifluoromethoxy)toluene
Hazard: Corrosive/Lachrymator
Formula:
C₈H₆BrF₃O Density: 1.5838 |
MDL Number:
MFCD00061271 CAS Number: 159689-88-0 UN: 3265 ADR: 8,III |
Specification Test Results
Density | 1.5838 |
ND20 | 1.4814 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...