(±)-2,5-Difluoromandelic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (±)-2,5-Difluorophenylglycolic acid

005297-5G 005297-25G
101778-212EA 32.31 USD
101778-212 101778-214
(±)-2,5-Difluoromandelic acid ≥97%
(±)-2,5-Difluoromandelic acid

Hazard: Irritant

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00061297
CAS Number: 207853-61-0

Order Now

Specification Test Results

Melting point 110-112°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR