2,2-Difluoroethyl acetate ≥97%
Supplier: Matrix Scientific
Synonyms:
2,2-Difluoroethylacetate
005271-25G
005271-5G
101778-198EA
312.31
USD
101778-198
101778-196
2,2-Difluoroethyl acetate ≥97%
2,2-Difluoroethyl acetate
Hazard: Flammable
Formula:
C₄H₆F₂O₂ MW: 124.09 g/mol Boiling Pt: 105 °C |
MDL Number:
MFCD00069133 CAS Number: 1550-44-3 |
Specification Test Results
Boiling point | 105° |
ND20 | 1.3542 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...