2,2-Difluoroethanol ≥95%

Supplier: Matrix Scientific

005270-25G 005270-100G
101778-192EA 75.38 USD
101778-192 101778-194
2,2-Difluoroethanol ≥95%
2,2-Difluoroethanol

Hazard: Flammable/Toxic

Formula: C₂H₄F₂O
MW: 82.05 g/mol
Boiling Pt: 95-96°C
Density: 1.296
MDL Number: MFCD00042248
CAS Number: 359-13-7
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Boiling point 95-96°
Density 1.296

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR