Ethyl chlorodifluoroacetate ≥99%

Supplier: Matrix Scientific

005265-25G 005265-100G
101778-180EA 30.77 USD
101778-180 101778-182
Ethyl chlorodifluoroacetate ≥99%
Ethyl chlorodifluoroacetate

TCA listed: T
Hazard: Flammable

Formula: C₄H₅ClF₂O₂
MW: 158.53 g/mol
Boiling Pt: 96…97 °C
Density: 1.252
MDL Number: MFCD00013662
CAS Number: 383-62-0
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Boiling point 96-97°
Density 1.252
ND20 1.3577

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR