3,5-Difluorobenzylamine ≥97%

Supplier: Matrix Scientific

005250-5G 005250-1G
101778-170EA 32.31 USD
101778-170 101778-168
3,5-Difluorobenzylamine ≥97%
3,5-Difluorobenzylamine

Hazard: Corrosive

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 184°C
Density: 1.21
Flash Pt: 73°C
MDL Number: MFCD00061244
CAS Number: 90390-27-5
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR