3,4-Difluorobenzylamine ≥97%

Supplier: Matrix Scientific

005249-25G 005249-100G
101778-166EA 73.85 USD
101778-166 102550-856
3,4-Difluorobenzylamine ≥97%
3,4-Difluorobenzylamine

Hazard: Corrosive

Formula: C₇H₇F₂N
Density: 1.21
MDL Number: MFCD00010145
CAS Number: 72235-53-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Density 1.21
ND20 1.493


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR