2,4-Difluorobenzylamine ≥97%

Supplier: Matrix Scientific

Synonyms: (2,4-Difluorophenyl)methanamine

005248-25G 005248-5G 005248-100G
101778-160EA 56.92 USD
101778-160 101778-158 101778-162
2,4-Difluorobenzylamine ≥97%
2,4-Difluorobenzylamine

Hazard: Corrosive

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 55…57 °C (8 mmHg)
Density: 1.204
MDL Number: MFCD00010142
CAS Number: 72235-52-0
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Boiling point 55-57°/8mm
Density 1.204
ND20 1.49

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR