2,6-Difluorobenzylamine ≥97%

Supplier: Matrix Scientific

005247-5G 005247-1G
101778-156EA 92.31 USD
101778-156 101778-154
2,6-Difluorobenzylamine ≥97%
2,6-Difluorobenzylamine

Hazard: Corrosive

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Density: 1.197
MDL Number: MFCD00010144
CAS Number: 69385-30-4
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Density 1.197
ND20 1.493

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR