3,5-Difluorobenzyl bromide ≥98%

Supplier: Matrix Scientific

Synonyms: α-Bromo-3,5-difluorotoluene

005244-25G 005244-100G
101778-150EA 115.38 USD
101778-150 101778-152
3,5-Difluorobenzyl bromide ≥98%
3,5-Difluorobenzyl bromide

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 65°C/5mm
Density: 1.6
MDL Number: MFCD00010304
CAS Number: 141776-91-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 65°/5mm
Density 1.6
ND20 1.521

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR