2,4-Difluorobenzyl bromide ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene

005242-100G 005242-25G
101778-148EA 167.69 USD
101778-148 101778-146
2,4-Difluorobenzyl bromide ≥95%
2,4-Difluorobenzyl bromide

Hazard: Flammable/Corrosive/Refrigerate/Lachrymator

Formula: C₇H₅BrF₂
Boiling Pt: 28°C/0.4mm
Density: 1.613
MDL Number: MFCD00011649
CAS Number: 23915-07-3
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Boiling point 28°/0.4mm
Density 1.613
ND20 1.525


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR